Learn More
3-(1-Naphthyl)acrylic acid, 98%
CAS: 13026-12-5 | C13H10O2 | 198.22 g/mol
Supplier: Thermo Scientific Chemicals A1542218
Description
3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
| 3-(1-Naphthyl)acrylic acid | |
| 210°C to 212°C | |
| MFCD00014317,MFCD00014317 | |
| 1868617 | |
| Slightly soluble in water. | |
| OC(=O)\C=C\C1=C2C=CC=CC2=CC=C1 | |
| 198.22 | |
| 198.22 |
| 13026-12-5 | |
| C13H10O2 | |
| 50 g | |
| naphthalene-1-acrylic acid, z-3-1-naphthyl acrylic acid, z-3-naphthalen-1-yl-acrylic acid, z-3-naphthalen-1-yl acrylic acid, 2z-3-naphthalen-1-yl prop-2-enoic acid | |
| WPXMLUUYWNHQOR-CMDGGOBGSA-N | |
| (Z)-3-naphthalen-1-ylprop-2-enoic acid | |
| 12158295 | |
| 98% |
Chemical Identifiers
| 13026-12-5 | |
| 198.22 | |
| WPXMLUUYWNHQOR-CMDGGOBGSA-N | |
| 12158295 |
| C13H10O2 | |
| MFCD00014317,MFCD00014317 | |
| naphthalene-1-acrylic acid, z-3-1-naphthyl acrylic acid, z-3-naphthalen-1-yl-acrylic acid, z-3-naphthalen-1-yl acrylic acid, 2z-3-naphthalen-1-yl prop-2-enoic acid | |
| OC(=O)\C=C\C1=C2C=CC=CC2=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 235-887-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only