missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)acrylic acid, 98%

Catalog No. p-7022578
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1542209 10 g
AAA1542218 50 g
AAA1542230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1542209 Supplier Thermo Scientific Chemicals Supplier No. A1542209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13026-12-5 | C13H10O2 | 198.22 g/mol

3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 13026-12-5
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00014317,MFCD00014317
InChI Key WPXMLUUYWNHQOR-CMDGGOBGSA-N
Synonym naphthalene-1-acrylic acid, z-3-1-naphthyl acrylic acid, z-3-naphthalen-1-yl-acrylic acid, z-3-naphthalen-1-yl acrylic acid, 2z-3-naphthalen-1-yl prop-2-enoic acid
PubChem CID 12158295
SMILES OC(=O)\C=C\C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 210°C to 212°C
Quantity 10 g
Beilstein 1868617
Solubility Information Slightly soluble in water.
IUPAC Name (2E)-3-(naphthalen-1-yl)prop-2-enoic acid
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3-(1-Naphthyl)acrylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-887-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only