missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-4-piperidinyl)indole, 97%

Catalog No. AAH63020MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63020MD 250 mg
AAH6302006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63020MD Supplier Thermo Scientific Chemicals Supplier No. H63020MD

CAS: 17403-07-5 | C14H18N2 | 214.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17403-07-5
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.31
MDL Number MFCD07776765
InChI Key KYSCKYJNMTUJPA-UHFFFAOYSA-N
PubChem CID 11206672
IUPAC Name 3-(1-methylpiperidin-4-yl)-1H-indole
SMILES CN1CCC(CC1)C1=CNC2=CC=CC=C12

Specifications

Color White to Yellow
Quantity 250 mg
Beilstein 1573327
Synonym 3-1-methyl-4-piperidinyl-1h-indole, 3-n-methylpiperidinyl indole, 3-1-methylpiperidin-4-yl-1h-indole, unii-7nsm99c49f, 3-1-methyl-4-piperidinyl indole, 1h-indole, 3-1-methyl-4-piperidinyl, naratriptan impurity a, usp naratriptan related compound a free base, naratriptan hydrochloride impurity, 3-1-methylpiperidin-4-yl-1h-indole-usp, 1h-indole,3-1-methyl-4-piperidinyl
Formula Weight 214.31
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(1-Methyl-4-piperidinyl)indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only