accessibility menu, dialog, popup

UserName

H3CNHN

CAS RN 17403-07-5

IUPAC Name: 3-(1-methylpiperidin-4-yl)-1H-indole
Synonyms: 3-(1-methylpiperidin-4-yl)-1H-indole
Molecular Weight (g/mol): 214.31
Molecular Formula: C14H18N2
InChi Key: KYSCKYJNMTUJPA-UHFFFAOYSA-N
SMILES: CN1CCC(CC1)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results