missing translation for 'onlineSavingsMsg'
Learn More

3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%

Catalog No. p-7030673
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3146503 1 g
AAH3146506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3146503 Supplier Thermo Scientific Chemicals Supplier No. H3146503
Only null left

CAS: 87151-66-4 | C9H14O | 138.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87151-66-4
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD12407081
InChI Key DJJNAQALDCPMOU-UHFFFAOYSA-N
Synonym 3-1,4-cyclohexadien-1-yl-1-propanol, 3-cyclohexa-1,4-dien-1-yl propan-1-ol, 3-1,4-cyclohexadienyl propan-1-ol, 3-3-hydroxypropyl-1,4-cyclohexadiene
PubChem CID 20402847
IUPAC Name 3-cyclohexa-1,4-dien-1-ylpropan-1-ol
SMILES C1C=CCC(=C1)CCCO

Specifications

Quantity 1 g
Sensitivity Air sensitive
Formula Weight 138.21
Percent Purity 97%
Chemical Name or Material 3-(1,4-Cyclohexadien-1-yl)-1-propanol
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only