missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Catalog No. p-3541034
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC58801CB 250mg Amber Glass Bottle
CC58801DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC58801CB Supplier Thermo Scientific Chemicals Supplier No. CC58801CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 934570-44-2
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.21
MDL Number MFCD09702391
InChI Key ZODQJPSGAHBVHU-UHFFFAOYSA-N
Synonym 3-1,3-dioxolan-2-yl thiophene-2-carboxylic acid, 3-1,3-dioxolan-2-yl-2-thiophenecarboxylic acid, 2-2-carboxythien-3-yl-1,3-dioxolane, 3-1,3-dioxolan-2-yl thiophene-2-carboxylicacid, 2-thiophenecarboxylicacid, 3-1,3-dioxolan-2-yl
PubChem CID 24229593
IUPAC Name 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylic acid
SMILES OC(=O)C1=C(C=CS1)C1OCCO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 200.21
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Dioxolan-2-yl)-2-thiophenecarboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.