missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one 98.0+%, TCI America™

Catalog No. B49835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49835G 5 g
B49831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49835G Supplier TCI America Supplier No. B49835G
Only null left

Chemical Identifiers

CAS 644-34-8
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00022990
InChI Key ATKADZVINWFQOE-SOFGYWHQSA-N
Synonym 3,4-(Methylenedioxy)chalcone
PubChem CID 5354492
IUPAC Name (E)-3-(1,3-benzodioxol-5-yl)-1-phenylprop-2-en-1-one
SMILES C1OC2=C(O1)C=C(C=C2)C=CC(=O)C3=CC=CC=C3

Specifications

Melting Point 123°C
Color Yellow
Quantity 5 g
Formula Weight 252.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one
RTECSNumber UD5572629
TSCA No