accessibility menu, dialog, popup

UserName

OOO(E)

CAS RN 644-34-8

IUPAC Name: (2E)-3-(2H-1,3-benzodioxol-5-yl)-1-phenylprop-2-en-1-one
Synonyms: (2E)-3-(2H-1,3-benzodioxol-5-yl)-1-phenylprop-2-en-1-one
Molecular Weight (g/mol): 252.27
Molecular Formula: C16H12O3
InChi Key: ATKADZVINWFQOE-SOFGYWHQSA-N
SMILES: O=C(\C=C\C1=CC=C2OCOC2=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results