missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1,2-Dimethyl-3-indolyl)-3-[4-(diethylamino)-2-methylphenyl]phthalide 98.0+%, TCI America™
Supplier: TCI America D320125G
Specifications
| 3-(1,2-Dimethyl-3-indolyl)-3-[4-(diethylamino)-2-methylphenyl]phthalide | |
| 217°C | |
| C29H30N2O2 | |
| LCGJEUAHECYSFW-UHFFFAOYSA-N | |
| 3-[4-(diethylamino)-2-methylphenyl]-3-(1,2-dimethylindol-3-yl)-2-benzofuran-1-one | |
| 22174174 | |
| ≥98.0% (HPLC,T) |
| 36499-49-7 | |
| Yellow | |
| 25 g | |
| CCN(CC)C1=CC(=C(C=C1)C2(C3=CC=CC=C3C(=O)O2)C4=C(N(C5=CC=CC=C54)C)C)C | |
| 438.571 | |
| 438.57 | |
| Crystalline Powder |
Chemical Identifiers
| 36499-49-7 | |
| 438.571 | |
| 22174174 | |
| CCN(CC)C1=CC(=C(C=C1)C2(C3=CC=CC=C3C(=O)O2)C4=C(N(C5=CC=CC=C54)C)C)C |
| C29H30N2O2 | |
| LCGJEUAHECYSFW-UHFFFAOYSA-N | |
| 3-[4-(diethylamino)-2-methylphenyl]-3-(1,2-dimethylindol-3-yl)-2-benzofuran-1-one |
Safety and Handling
TSCA : No