missing translation for 'onlineSavingsMsg'
Learn More

3-(1,2-Dimethyl-3-indolyl)-3-[4-(diethylamino)-2-methylphenyl]phthalide 98.0+%, TCI America™

Catalog No. p-7127621
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32015G 5 g
D320125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32015G Supplier TCI America Supplier No. D32015G
Only null left

Chemical Identifiers

CAS 36499-49-7
Molecular Formula C29H30N2O2
Molecular Weight (g/mol) 438.571
InChI Key LCGJEUAHECYSFW-UHFFFAOYSA-N
PubChem CID 22174174
IUPAC Name 3-[4-(diethylamino)-2-methylphenyl]-3-(1,2-dimethylindol-3-yl)-2-benzofuran-1-one
SMILES CCN(CC)C1=CC(=C(C=C1)C2(C3=CC=CC=C3C(=O)O2)C4=C(N(C5=CC=CC=C54)C)C)C

Specifications

Melting Point 217°C
Color Yellow
Quantity 5 g
Formula Weight 438.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1,2-Dimethyl-3-indolyl)-3-[4-(diethylamino)-2-methylphenyl]phthalide