missing translation for 'onlineSavingsMsg'
Learn More

2xYT Broth (Powder/Molecular Genetics), Fisher BioReagents

Catalog No. BP24672
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP24672 2000 g Poly Bottle
BP2467500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP24672 Supplier Fisher BioReagents Supplier No. BP2467 2
Get Up to $1,800 in Chemicals with Purchase
View all promotions

For use in molecular genetics

Formulation Usage: 31.0g/L

Specifications

Content And Storage <4°C
Additive 2xyt Broth
Form Powder
Product Type Bacterial Culture
Formulation 31g/L
For Use With (Application) Molecular Genetics
Packaging Type Poly Bottle
Quantity 2000 g