missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-(+)-Glycidyl p-Toluenesulfonate 98.0+%, TCI America™
Supplier: TCI America T161225G
Specifications
| (2S)-(+)-Glycidyl p-Toluenesulfonate | |
| 48°C | |
| C10H12O4S | |
| 25 g | |
| s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s | |
| CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 | |
| 228.262 | |
| 228.26 | |
| Crystalline Powder |
| 70987-78-9 | |
| White | |
| MFCD00064489 | |
| 3077 | |
| NOQXXYIGRPAZJC-VIFPVBQESA-N | |
| [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate | |
| 153296 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 70987-78-9 | |
| 228.262 | |
| NOQXXYIGRPAZJC-VIFPVBQESA-N | |
| 153296 | |
| CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 |
| C10H12O4S | |
| MFCD00064489 | |
| s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s | |
| [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate |
Safety and Handling
TSCA : No