accessibility menu, dialog, popup

UserName

CAS RN 70987-78-9

CH3SOOO(S)O

CAS RN 70987-78-9

IUPAC Name: [(2S)-oxiran-2-yl]methyl 4-methylbenzene-1-sulfonate
Synonyms: (2S)-oxiran-2-ylmethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 228.26
Molecular Formula: C10H12O4S
InChi Key: NOQXXYIGRPAZJC-VIFPVBQESA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H]1CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results