missing translation for 'onlineSavingsMsg'
Learn More

(2S)-(+)-Glycidyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. T16125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16125G 5 g
T161225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16125G Supplier TCI America Supplier No. T16125G
Only null left

Chemical Identifiers

CAS 70987-78-9
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD00064489
InChI Key NOQXXYIGRPAZJC-VIFPVBQESA-N
Synonym s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s
PubChem CID 153296
IUPAC Name [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2

Specifications

Melting Point 48°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 228.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S)-(+)-Glycidyl p-Toluenesulfonate