missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone 97.0+%, TCI America™
$218.10 - $868.65
Chemical Identifiers
| CAS | 346440-54-8 |
|---|---|
| Molecular Formula | C15H22N2O |
| Molecular Weight (g/mol) | 246.35 |
| MDL Number | MFCD03426982 |
| InChI Key | SKHPYKHVYFTIOI-UHFFFAOYNA-N |
| Synonym | (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone |
| PubChem CID | 10309834 |
| IUPAC Name | 5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one |
| SMILES | CN1C(NC(CC2=CC=CC=C2)C1=O)C(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4138200MG
|
TCI America
B4138200MG |
200 mg |
Each for $218.10
|
|
|||||
|
B41381G
|
TCI America
B41381G |
1 g |
Each for $868.65
|
|
|||||
Chemical Identifiers
| 346440-54-8 | |
| 246.35 | |
| SKHPYKHVYFTIOI-UHFFFAOYNA-N | |
| 10309834 | |
| CN1C(NC(CC2=CC=CC=C2)C1=O)C(C)(C)C |
| C15H22N2O | |
| MFCD03426982 | |
| (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone | |
| 5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one |
Specifications
| 346440-54-8 | |
| C15H22N2O | |
| 200 mg | |
| SKHPYKHVYFTIOI-UHFFFAOYNA-N | |
| 5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one | |
| 10309834 | |
| ≥97.0% (GC) | |
| (2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone |
| 100°C | |
| MFCD03426982 | |
| (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone | |
| CN1C(NC(CC2=CC=CC=C2)C1=O)C(C)(C)C | |
| 246.35 | |
| 246.35 | |
| Crystalline Powder |
Safety and Handling
TSCA : No