CAS RN 346440-54-8
IUPAC Name:
5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one
Synonyms:
5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one
Molecular Weight (g/mol):
246.35
Molecular Formula:
C15H22N2O
InChi Key:
SKHPYKHVYFTIOI-UHFFFAOYNA-N
SMILES:
CN1C(NC(CC2=CC=CC=C2)C1=O)C(C)(C)C
1
–
1
of
1
results
1
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone 97.0+%, TCI America™
CAS: 346440-54-8 Molecular Formula: C15H22N2O Molecular Weight (g/mol): 246.35 MDL Number: MFCD03426982 InChI Key: SKHPYKHVYFTIOI-UHFFFAOYNA-N Synonym: (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone PubChem CID: 10309834 IUPAC Name: 5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one SMILES: CN1C(NC(CC2=CC=CC=C2)C1=O)C(C)(C)C