missing translation for 'onlineSavingsMsg'
Learn More

(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone 97.0+%, TCI America™

Catalog No. B4138200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4138200MG 200 mg
B41381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4138200MG Supplier TCI America Supplier No. B4138200MG
Only null left

Chemical Identifiers

CAS 346440-54-8
Molecular Formula C15H22N2O
Molecular Weight (g/mol) 246.35
MDL Number MFCD03426982
InChI Key SKHPYKHVYFTIOI-UHFFFAOYNA-N
Synonym (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone
PubChem CID 10309834
IUPAC Name 5-benzyl-2-tert-butyl-3-methylimidazolidin-4-one
SMILES CN1C(NC(CC2=CC=CC=C2)C1=O)C(C)(C)C

Specifications

Melting Point 100°C
Quantity 200 mg
Formula Weight 246.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone