missing translation for 'onlineSavingsMsg'
Learn More

(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane, 99%, Thermo Scientific Chemicals

Catalog No. p-7050052
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33668MD 250 mg
AA3366803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33668MD Supplier Thermo Scientific Chemicals Supplier No. 033668MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77876-39-2
Molecular Formula C29H30P2
Molecular Weight (g/mol) 440.51
MDL Number MFCD00058923
InChI Key CTYPJIUQROQJBG-ISIPKWIHNA-N
Synonym 2s,4s---2,4-bis diphenylphosphino pentane, s,s-bdpp, 2s,4s-pentane-2,4-diylbis diphenylphosphine, phosphine, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenyl, s,s-2,4-bis diphenylphosphino pentane, 2s,4s-2,4-bis diphenylphosphino pentane, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenylphosphine, 2s,4s-4-diphenylphosphanylpentan-2-yl-diphenylphosphane, 2s,4s-4-diphenylphosphanyl pentan-2-yl diphenylphosphane
PubChem CID 2734567
SMILES C[C@@H](C[C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 250 mg
IUPAC Name [(2S,4S)-4-diphenylphosphanylpentan-2-yl]-diphenylphosphane
Formula Weight 440.5
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material (2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only