accessibility menu, dialog, popup

UserName

CAS RN 77876-39-2

IUPAC Name: [(2S,4S)-4-(diphenylphosphanyl)pentan-2-yl]diphenylphosphane
Synonyms: [(2S,4S)-4-(diphenylphosphanyl)pentan-2-yl]diphenylphosphane
Molecular Weight (g/mol): 440.51
Molecular Formula: C29H30P2
InChi Key: CTYPJIUQROQJBG-ISIPKWIHNA-N
SMILES: C[C@@H](C[C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results