Learn More
(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
CAS: 61478-28-2 | C34H37NO2P2 | 553.62 g/mol
Supplier: Thermo Scientific Chemicals H2732103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine | |
| 100°C to 103°C | |
| MFCD01631275 | |
| 2s,4s---n-boc-4-diphenylphosphino-2-diphenylphosphinomethyl-pyrrolidine, bppm, s,s-bppm, 2s,4s-1-tert-butoxycarbonyl-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---2-diphenylphosphinomethyl-4-diphenylphosphino-n-t-butoxycarbonyl pyrrolidine, 2s,4s-tert-butyl 4-diphenylphosphino-2-diphenylphosphino methyl pyrrolidine-1-carboxylate, tert-butyl 2s,4s-4-diphenylphosphanyl-2-diphenylphosphanyl methyl pyrrolidine-1-carboxylate, 2s,4s-1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine | |
| CC(C)(C)OC(=O)N1C[C@H](C[C@H]1CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 553.62 | |
| 553.62 |
| 61478-28-2 | |
| C34H37NO2P2 | |
| 1 g | |
| BFMKBYZEJOQYIM-FEHSQGEHNA-N | |
| tert-butyl (2S,4S)-4-diphenylphosphanyl-2-(diphenylphosphanylmethyl)pyrrolidine-1-carboxylate | |
| 10209545 |
Chemical Identifiers
| 61478-28-2 | |
| 553.62 | |
| BFMKBYZEJOQYIM-FEHSQGEHNA-N | |
| 10209545 | |
| CC(C)(C)OC(=O)N1C[C@H](C[C@H]1CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C34H37NO2P2 | |
| MFCD01631275 | |
| 2s,4s---n-boc-4-diphenylphosphino-2-diphenylphosphinomethyl-pyrrolidine, bppm, s,s-bppm, 2s,4s-1-tert-butoxycarbonyl-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---2-diphenylphosphinomethyl-4-diphenylphosphino-n-t-butoxycarbonyl pyrrolidine, 2s,4s-tert-butyl 4-diphenylphosphino-2-diphenylphosphino methyl pyrrolidine-1-carboxylate, tert-butyl 2s,4s-4-diphenylphosphanyl-2-diphenylphosphanyl methyl pyrrolidine-1-carboxylate, 2s,4s-1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine | |
| tert-butyl (2S,4S)-4-diphenylphosphanyl-2-(diphenylphosphanylmethyl)pyrrolidine-1-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only