missing translation for 'onlineSavingsMsg'
Learn More

(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine

Catalog No. p-7029812
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27321MD 250 mg
AAH2732103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27321MD Supplier Thermo Scientific Chemicals Supplier No. H27321MD
Only null left

CAS: 61478-28-2 | C34H37NO2P2 | 553.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61478-28-2
Molecular Formula C34H37NO2P2
Molecular Weight (g/mol) 553.62
MDL Number MFCD01631275
InChI Key BFMKBYZEJOQYIM-FEHSQGEHNA-N
Synonym 2s,4s---n-boc-4-diphenylphosphino-2-diphenylphosphinomethyl-pyrrolidine, bppm, s,s-bppm, 2s,4s-1-tert-butoxycarbonyl-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---2-diphenylphosphinomethyl-4-diphenylphosphino-n-t-butoxycarbonyl pyrrolidine, 2s,4s-tert-butyl 4-diphenylphosphino-2-diphenylphosphino methyl pyrrolidine-1-carboxylate, tert-butyl 2s,4s-4-diphenylphosphanyl-2-diphenylphosphanyl methyl pyrrolidine-1-carboxylate, 2s,4s-1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine
PubChem CID 10209545
IUPAC Name tert-butyl (2S,4S)-4-diphenylphosphanyl-2-(diphenylphosphanylmethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C[C@H](C[C@H]1CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 100°C to 103°C
Quantity 250 mg
Formula Weight 553.62
Chemical Name or Material (2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only