missing translation for 'onlineSavingsMsg'
Learn More

(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96.0+%, TCI America™

Catalog No. p-7123084
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36621G 1 g
B36625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36621G Supplier TCI America Supplier No. B36621G
Only null left

Chemical Identifiers

CAS 61478-26-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD02094386
InChI Key UFJNFQNQLMGUTQ-JGVFFNPUSA-N
PubChem CID 11275979
IUPAC Name tert-butyl (2S,4R)-4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1CO)O

Specifications

Melting Point 84°C
Quantity 1 g
Synonym n-boc-trans-4-hydroxy-l-prolinol, boc-hyp-ol, 2s,4r-tert-butyl 4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s,4r-4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, boc-trans-4-hydroxy-l-prolinol, 2s,4r-1-tert-butoxycarbonyl-4-hydroxy-2-hydroxymethyl pyrrolidine, 2s,4r-1-boc-4-hydroxy-2-hydroxymethyl pyrrolidine, n-boc-2s,4r-4-hydroxy-2-hydroxymethylpyrrolidine, 2s,4r-tert-butyl4-hydroxy-2-hydroxymethyl pyrrolidine-1-carboxylate, trans-4-hydroxy-l-prolinol, n-boc protected
Formula Weight 217.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine