missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%

Catalog No. AC290032500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290032500 250 mg Glass bottle
AC290030010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC290032500 Supplier Thermo Scientific Chemicals Supplier No. 290032500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104195-80-4 | C5H11NO3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104195-80-4
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
InChI Key FYCWLJLGIAUCCL-IMJSIDKUSA-N
Synonym 2s,3s-2-amino-3-methoxybutanoic acid, 2s,3s-2-amino-3-methoxy-butanoic acid, allo-o-methyl-l-thr, l-allo-o-methylthreonine, 2s,3s-2-amino-3-methoxybutanoic acid
PubChem CID 151207
IUPAC Name (2S,3S)-2-amino-3-methoxybutanoic acid
SMILES CC(C(C(=O)O)N)OC

Specifications

Melting Point 220.0°C to 224.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH(OCH3)CH(NH2)COOH
Quantity 250 mg
Specific Rotation Condition + 27.50 (25.00°C c=0.6,6N HCl)
Specific Rotation + 27.50
Solubility Information Solubility in water: soluble in water.
Formula Weight 133.15
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material (2S, 3S)-2-Amino-3-methoxybutanoic acid
Show More Show Less

RUO – Research Use Only