CAS RN 104195-80-4
CAS RN 104195-80-4
IUPAC Name:
(2S,3S)-2-azaniumyl-3-methoxybutanoate
Synonyms:
(2S,3S)-2-ammonio-3-methoxybutanoate
Molecular Weight (g/mol):
133.15
Molecular Formula:
C5H11NO3
InChi Key:
FYCWLJLGIAUCCL-IMJSIDKUSA-N
SMILES:
CO[C@@H](C)[C@H]([NH3+])C([O-])=O
1
–
2
of
2
results
1
(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%
CAS: 104195-80-4 Molecular Formula: C5H11NO3 Molecular Weight (g/mol): 133.15 InChI Key: FYCWLJLGIAUCCL-IMJSIDKUSA-N Synonym: 2s,3s-2-amino-3-methoxybutanoic acid,2s,3s-2-amino-3-methoxy-butanoic acid,allo-o-methyl-l-thr,l-allo-o-methylthreonine, 2s,3s-2-amino-3-methoxybutanoic acid PubChem CID: 151207 IUPAC Name: (2S,3S)-2-amino-3-methoxybutanoic acid SMILES: CC(C(C(=O)O)N)OC