missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one 96.0+%, TCI America™

Catalog No. D36615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36615G 5 g
D366125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36615G Supplier TCI America Supplier No. D36615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42399-49-5
Molecular Formula C16H15NO3S
Molecular Weight (g/mol) 301.36
MDL Number MFCD01321316
InChI Key LHBHZALHFIQJGJ-CABCVRRESA-N
Synonym azepin-4-one, 2s,3s-+-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, unii-6rn4n3305m, 2s-cis-+-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, 2s,3s-3-hydroxy-2-4-methoxyphenyl-2,3-dihydro-1,5-benzothiazepin-4 5h-one, 2s,3s-3-hydroxy-2-4-methoxyphenyl-3,5-dihydro-2h-1,5-benzothiazepin-4-one, 2s-cis-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, 3-hydroxy-2-4-methoxyphenyl-2,3-dihydro-1,5-benzothiazepin-4 5h-one, 2s,3s-2,3-dihydro-3-hydroxy-2-4-methoxyphenyl-1,5-benzothiazepin-4 5h-one, diltiazem impurity e
PubChem CID 2733673
IUPAC Name (2S,3S)-3-hydroxy-2-(4-methoxyphenyl)-3,5-dihydro-2H-1,5-benzothiazepin-4-one
SMILES COC1=CC=C(C=C1)C2C(C(=O)NC3=CC=CC=C3S2)O

Specifications

Melting Point 204°C
Color Yellow
Quantity 5 g
Formula Weight 301.36
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one
EINECSNumber (5)-3876
TSCA No