accessibility menu, dialog, popup

UserName

CAS RN 42399-49-5

IUPAC Name: (2S,3S)-3-hydroxy-2-(4-methoxyphenyl)-2,3,4,5-tetrahydro-1,5-benzothiazepin-4-one
Synonyms: (2S,3S)-3-hydroxy-2-(4-methoxyphenyl)-3,5-dihydro-2H-1,5-benzothiazepin-4-one
Molecular Weight (g/mol): 301.36
Molecular Formula: C16H15NO3S
InChi Key: LHBHZALHFIQJGJ-CABCVRRESA-N
SMILES: COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2NC(=O)[C@@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results