missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene 98.0+%, TCI America™

Catalog No. b3450100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3450100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3450100MG Supplier TCI America Supplier No. B3450100MG

Chemical Identifiers

CAS 71042-54-1
Molecular Formula C31H28P2
Molecular Weight (g/mol) 462.513
MDL Number MFCD00085364
InChI Key CDJHPMXMJUCLPA-WDHYYQACSA-N
Synonym 2s,3s-+-2,3-bis diphenylphosphino bicyclo 2.2.1 hept-5-ene, 2s,3s-norphos, 2s,3s-+-norphos, 2s,3s-3-diphenylphosphanyl bicyclo 2.2.1 hept-5-en-2-yl diphenylphosphane, phosphine,1,1'-1r,2s,3s,4s-bicyclo 2.2.1 hept-5-ene-2,3-diylbis 1,1-diphenyl
PubChem CID 133126730
IUPAC Name [(1R,2S,3S)-2-diphenylphosphanyl-3-bicyclo[2.2.1]hept-5-enyl]-diphenylphosphane
SMILES C1C2C=CC1C(C2P(C3=CC=CC=C3)C4=CC=CC=C4)P(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 130°C
Color White
Quantity 100 mg
Formula Weight 462.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene