CAS RN 71042-54-1
IUPAC Name:
[3-(diphenylphosphanyl)bicyclo[2.2.1]hept-5-en-2-yl]diphenylphosphane
Synonyms:
[3-(diphenylphosphanyl)bicyclo[2.2.1]hept-5-en-2-yl]diphenylphosphane
Molecular Weight (g/mol):
462.51
Molecular Formula:
C31H28P2
InChi Key:
CDJHPMXMJUCLPA-UHFFFAOYNA-N
SMILES:
C1C2C=CC1C(C2P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(2S,3S)-(+)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene 98.0+%, TCI America™
CAS: 71042-54-1 Molecular Formula: C31H28P2 Molecular Weight (g/mol): 462.513 MDL Number: MFCD00085364 InChI Key: CDJHPMXMJUCLPA-WDHYYQACSA-N Synonym: 2s,3s-+-2,3-bis diphenylphosphino bicyclo 2.2.1 hept-5-ene,2s,3s-norphos,2s,3s-+-norphos,2s,3s-3-diphenylphosphanyl bicyclo 2.2.1 hept-5-en-2-yl diphenylphosphane,phosphine,1,1'-1r,2s,3s,4s-bicyclo 2.2.1 hept-5-ene-2,3-diylbis 1,1-diphenyl PubChem CID: 133126730 IUPAC Name: [(1R,2S,3S)-2-diphenylphosphanyl-3-bicyclo[2.2.1]hept-5-enyl]-diphenylphosphane SMILES: C1C2C=CC1C(C2P(C3=CC=CC=C3)C4=CC=CC=C4)P(C5=CC=CC=C5)C6=CC=CC=C6