missing translation for 'onlineSavingsMsg'
Learn More

(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, 97%

Catalog No. p-7035070
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52775MD 250 mg
AAH5277503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52775MD Supplier Thermo Scientific Chemicals Supplier No. H52775MD

CAS: 62023-65-8 | C15H21NO5 | 295.34 g/mol

(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, is used as a novel protected amino acid for the preparation of bestatin and analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, is used as a novel protected amino acid for the preparation of bestatin and analogs.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 62023-65-8
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00057841
InChI Key BHTRKISIDQZUQX-UHFFFAOYNA-N
Synonym 2s,3r-3-boc-amino-2-hydroxy-4-phenylbutyric acid, 2s,3r-3-tert-butoxycarbonyl amino-2-hydroxy-4-phenylbutanoic acid, n-boc-2s,3r-2-hydroxy-3-amino-4-phenylbutanoic acid, boc-2s,3r-ahpa, boc-2s,3r-3-amino-2-hydroxy-4-phenylbutyric acid, n-boc-2s,3r-2-hydroxy-3-amino-4-phenylbutanoic a, 2s,3r-2-hydroxy-3-tert-butoxycarbonylamino-4-phenylbutanoic acid, 2s,3r-3-boc-amino-2-hydroxy-4-phenylbutyric acid tlc
PubChem CID 10756280
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(O)C(O)=O

Specifications

Beilstein 5909790
Solubility Information Insoluble in water. Soluble in organic solvents.
IUPAC Name (2S,3R)-2-hydroxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
Quantity 250 mg
Formula Weight 295.34
Percent Purity 97%
Chemical Name or Material (2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only