accessibility menu, dialog, popup

UserName

CAS RN 62023-65-8

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-4-phenylbutanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-2-hydroxy-4-phenylbutanoic acid
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: BHTRKISIDQZUQX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)