missing translation for 'onlineSavingsMsg'
Learn More

(2S,3aS,7aS)-Octahydroindole-2-carboxylic acid, 98%

Catalog No. AAH2789406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2789406 5 g
AAH2789403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2789406 Supplier Thermo Scientific Chemicals Supplier No. H2789406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80875-98-5 | C9H15NO2 | 169.224 g/mol

An intermediate in the synthesis of Perindopril. The completely diastereoselective α-alkylation of oxazolidinone provides a very convenient and concise route to enantiopure α-tetrasubstituted derivatives of this Oic stereoisomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An intermediate in the synthesis of Perindopril. The completely diastereoselective α-alkylation of oxazolidinone provides a very convenient and concise route to enantiopure α-tetrasubstituted derivatives of this Oic stereoisomer.

Solubility
Soluble in methanol and water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C.

Chemical Identifiers

CAS 80875-98-5
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.224
MDL Number MFCD07782125
InChI Key CQYBNXGHMBNGCG-FXQIFTODSA-N
Synonym 2s,3as,7as-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-octahydroindole-2-carboxylic acid, 2s,3as,7as-2-carboxyoctahydroindole, h-oic-oh, l-octahydroindole-2-carboxylic acid, unii-9id44u804i, 2-carboxyoctahydroindole, 2s,3as,7as, 1h-indole-2-carboxylic acid, octahydro-, 2s,3as,7as, 2s,3as,7as-2-carboxyperhydroindole, 2s,3as,7as-perhydroindole-2-carboxylic acid
PubChem CID 7408452
IUPAC Name (2S,3aS,7aS)-2,3,3a,4,5,6,7,7a-octahydro-1H-indole-2-carboxylic acid
SMILES C1CCC2C(C1)CC(N2)C(=O)O

Specifications

Melting Point 266°C (decomposition)
Quantity 5 g
Solubility Information Soluble in methanol and water.
Optical Rotation −50 (c=1 in Methanol)
Formula Weight 169.22
Percent Purity 98%
Chemical Name or Material (2S,3aS,7aS)-Octahydroindole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only