accessibility menu, dialog, popup

UserName

CAS RN 80875-98-5

IUPAC Name: octahydro-1H-indole-2-carboxylic acid
Synonyms: octahydro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 169.22
Molecular Formula: C9H15NO2
InChi Key: CQYBNXGHMBNGCG-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2CCCCC2N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)