missing translation for 'onlineSavingsMsg'
Learn More

(2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134299
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03701G 1 g
O03705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03701G Supplier TCI America Supplier No. O03701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80875-98-5
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.224
MDL Number MFCD07782125
InChI Key CQYBNXGHMBNGCG-FXQIFTODSA-N
Synonym 2s,3as,7as-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-octahydroindole-2-carboxylic acid, 2s,3as,7as-2-carboxyoctahydroindole, h-oic-oh, l-octahydroindole-2-carboxylic acid, unii-9id44u804i, 2-carboxyoctahydroindole, 2s,3as,7as, 1h-indole-2-carboxylic acid, octahydro-, 2s,3as,7as, 2s,3as,7as-2-carboxyperhydroindole, 2s,3as,7as-perhydroindole-2-carboxylic acid
PubChem CID 7408452
IUPAC Name (2S,3aS,7aS)-2,3,3a,4,5,6,7,7a-octahydro-1H-indole-2-carboxylic acid
SMILES C1CCC2C(C1)CC(N2)C(=O)O

Specifications

Melting Point 270°C
Color White
Quantity 1 g
Formula Weight 169.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic Acid