missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS,3SR)-2-Methylisocitric acid sodium salt ≥95.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 71183-66-9 |
| Linear Formula | C7H10O7 · xNa+ |
| Type | Organic Standard |
| Quantity | 50 mg |
| Formula Weight | 206.15 (free acid basis) |
| Percent Purity | ≥95% (GC) |