missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,5R)-2,5-Diphenylpyrrolidine 95.0+%, TCI America™
Supplier: TCI America D3185100MG
Specifications
| (2R,5R)-2,5-Diphenylpyrrolidine | |
| 50°C | |
| C16H17N | |
| 3077 | |
| C1CC(NC1C2=CC=CC=C2)C3=CC=CC=C3 | |
| 223.319 | |
| 223.32 | |
| Crystalline Powder |
| 155155-73-0 | |
| White-Yellow | |
| 100 mg | |
| NAOGHMNKMJMMNQ-HZPDHXFCSA-N | |
| (2R,5R)-2,5-diphenylpyrrolidine | |
| 10966132 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 155155-73-0 | |
| 223.319 | |
| 10966132 | |
| C1CC(NC1C2=CC=CC=C2)C3=CC=CC=C3 |
| C16H17N | |
| NAOGHMNKMJMMNQ-HZPDHXFCSA-N | |
| (2R,5R)-2,5-diphenylpyrrolidine |
Safety and Handling
TSCA : No