accessibility menu, dialog, popup

UserName

CAS RN 155155-73-0

HN

CAS RN 155155-73-0

IUPAC Name: 2,5-diphenylpyrrolidine
Synonyms: 2,5-diphenylpyrrolidine
Molecular Weight (g/mol): 223.32
Molecular Formula: C16H17N
InChi Key: NAOGHMNKMJMMNQ-UHFFFAOYNA-N
SMILES: C1CC(NC1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results