missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R,5S,6S)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane 98.0+%, TCI America™

Catalog No. B3695200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3695200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3695200MG Supplier TCI America Supplier No. B3695200MG
Only null left

Chemical Identifiers

CAS 173371-55-6
Molecular Formula C10H20O6
Molecular Weight (g/mol) 236.26
MDL Number MFCD20265640
InChI Key RAIJJZVROJTPEU-UHFFFAOYNA-N
Synonym (2S,3S,5R,6R)-5,6-Dimethoxy-5,6-dimethyl-1,4-dioxane-2,3-dimethanol
PubChem CID 10561775
IUPAC Name [3-(hydroxymethyl)-5,6-dimethoxy-5,6-dimethyl-1,4-dioxan-2-yl]methanol
SMILES COC1(C)OC(CO)C(CO)OC1(C)OC

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 200 mg
Physical Form Crystal-Powder at 20°C
Formula Weight 236.26
Percent Purity ≥98.0% (GC)
Chemical Name or Material (2R,3R,5S,6S)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane