CAS RN 173371-55-6
IUPAC Name:
[3-(hydroxymethyl)-5,6-dimethoxy-5,6-dimethyl-1,4-dioxan-2-yl]methanol
Synonyms:
[3-(hydroxymethyl)-5,6-dimethoxy-5,6-dimethyl-1,4-dioxan-2-yl]methanol
Molecular Weight (g/mol):
236.26
Molecular Formula:
C10H20O6
InChi Key:
RAIJJZVROJTPEU-UHFFFAOYNA-N
SMILES:
COC1(C)OC(CO)C(CO)OC1(C)OC
1
–
1
of
1
results
1
(2R,3R,5S,6S)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane 98.0+%, TCI America™
CAS: 173371-55-6 Molecular Formula: C10H20O6 Molecular Weight (g/mol): 236.26 MDL Number: MFCD20265640 InChI Key: RAIJJZVROJTPEU-UHFFFAOYNA-N Synonym: (2S,3S,5R,6R)-5,6-Dimethoxy-5,6-dimethyl-1,4-dioxane-2,3-dimethanol PubChem CID: 10561775 IUPAC Name: [3-(hydroxymethyl)-5,6-dimethoxy-5,6-dimethyl-1,4-dioxan-2-yl]methanol SMILES: COC1(C)OC(CO)C(CO)OC1(C)OC