missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2H-1,4-Benzothiazin-3(4H)-one 98.0+%, TCI America™
Supplier: TCI America B455425G
Specifications
| 2H-1,4-Benzothiazin-3(4H)-one | |
| 180°C | |
| C8H7NOS | |
| 25 g | |
| GTFMIJNXNMDHAB-UHFFFAOYSA-N | |
| 4H-1,4-benzothiazin-3-one | |
| 21396 | |
| ≥98.0% (GC) |
| 5325-20-2 | |
| White-Yellow | |
| MFCD00006698 | |
| 2h-1,4-benzothiazin-3 4h-one, 2h-benzo b 1,4 thiazin-3 4h-one, 2h 1,4-benzothiazin-3 4h-one, 2,4-dihydro-1,4-benzothiazin-3-one, 1,4-benzothiazin-3 4h-one, gnf-pf-3697, 4h-benzo 1,4 thiazin-3-one, 3,4-dihydro-2h-1,4-benzothiazin-3-one, 3,4-dihydro-3-oxo-2h-1,4-benzothiazine, 2h,4h-benzo e 1,4-thiazin-3-one | |
| C1C(=O)NC2=CC=CC=C2S1 | |
| 165.21 | |
| 165.21 | |
| Crystalline Powder |
Chemical Identifiers
| 5325-20-2 | |
| 165.21 | |
| GTFMIJNXNMDHAB-UHFFFAOYSA-N | |
| 21396 | |
| C1C(=O)NC2=CC=CC=C2S1 |
| C8H7NOS | |
| MFCD00006698 | |
| 2h-1,4-benzothiazin-3 4h-one, 2h-benzo b 1,4 thiazin-3 4h-one, 2h 1,4-benzothiazin-3 4h-one, 2,4-dihydro-1,4-benzothiazin-3-one, 1,4-benzothiazin-3 4h-one, gnf-pf-3697, 4h-benzo 1,4 thiazin-3-one, 3,4-dihydro-2h-1,4-benzothiazin-3-one, 3,4-dihydro-3-oxo-2h-1,4-benzothiazine, 2h,4h-benzo e 1,4-thiazin-3-one | |
| 4H-1,4-benzothiazin-3-one |
Safety and Handling
TSCA : No