missing translation for 'onlineSavingsMsg'
Learn More

2H-1,4-Benzothiazin-3(4H)-one 98.0+%, TCI America™

Catalog No. p-7123612
Change view
Click to view available options
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
B45541G 1 g
B45545G 5 g
B455425G 25 g
3 options

Catalog No. B45541G

Supplier: TCI America B45541G

Only null left

Chemical Identifiers

CAS 5325-20-2
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00006698
InChI Key GTFMIJNXNMDHAB-UHFFFAOYSA-N
Synonym 2h-1,4-benzothiazin-3 4h-one, 2h-benzo b 1,4 thiazin-3 4h-one, 2h 1,4-benzothiazin-3 4h-one, 2,4-dihydro-1,4-benzothiazin-3-one, 1,4-benzothiazin-3 4h-one, gnf-pf-3697, 4h-benzo 1,4 thiazin-3-one, 3,4-dihydro-2h-1,4-benzothiazin-3-one, 3,4-dihydro-3-oxo-2h-1,4-benzothiazine, 2h,4h-benzo e 1,4-thiazin-3-one
PubChem CID 21396
IUPAC Name 4H-1,4-benzothiazin-3-one
SMILES C1C(=O)NC2=CC=CC=C2S1

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 1 g
Formula Weight 165.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2H-1,4-Benzothiazin-3(4H)-one