missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ 293 Whole Cell Lysate
Description
The 293 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, SDS-Page.
Specifications
Specifications
| Quantity | 0.5 mg |
| Storage Buffer | 1X SDS-PAGE Sample Buffer [0.05mM Tris HCl, 2% SDS, 10% Glycerol, 0.1% bromophenol blue, pH 6.8] |
| Purity | Whole antisera |
| Test Specificity | 293 whole cell extract lysate |
| Cellular Fraction | Whole |
| Host Species | Human |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| For Use With (Application) | SDS-Page, Western Blot |
For Research Use Only