missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, Granular, Technical, Spectrum™ Chemical

Catalog No. 18612613
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612613 50 kg Poly Bottle
18612611 2.5 kg Poly Bottle
18612612 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612613 Supplier Spectrum Chemical Mfg Cor Supplier No. P128350KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7778-50-9

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 50 kg
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Potassium Dichromate, Granular

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.