missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol Methyl Ether, 99%, Spectrum™ Chemical
Specifications
Specifications
| Acidity | 0.0001 |
| CAS Max % | 1 |
| Boiling Point | 190°C/374°F |
| Packaging | Amber Glass Bottle |
| Quantity | 20 L |
| Specific Gravity | 0.945 - 0.957 |
| Percent Purity | 0.99 |
| Grade | Ungraded |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Dipropylene Glycol Methyl Ether |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.