missing translation for 'onlineSavingsMsg'
Learn More
Learn More
25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane 90.0+%, TCI America™
$83.89 - $249.78
Chemical Identifiers
| CAS | 869718-80-9 |
|---|---|
| Molecular Formula | C17H35N3O8 |
| Molecular Weight (g/mol) | 409.48 |
| MDL Number | MFCD13184962 |
| InChI Key | ANQOCZRUHGJYCX-UHFFFAOYSA-N |
| PubChem CID | 49800635 |
| IUPAC Name | 1-azido-2-[2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethane |
| SMILES | COCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A272725MG
|
TCI America
A272725MG |
25 mg |
Each for $83.89
|
|
|||||
|
A2727100MG
|
TCI America
A2727100MG |
100 mg |
Each for $249.78
|
|
|||||
Chemical Identifiers
| 869718-80-9 | |
| 409.48 | |
| ANQOCZRUHGJYCX-UHFFFAOYSA-N | |
| 1-azido-2-[2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethane |
| C17H35N3O8 | |
| MFCD13184962 | |
| 49800635 | |
| COCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-] |
Specifications
| 869718-80-9 | |
| C17H35N3O8 | |
| 25 mg | |
| ANQOCZRUHGJYCX-UHFFFAOYSA-N | |
| 1-azido-2-[2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethane | |
| 49800635 | |
| ≥90.0% (HPLC) | |
| 25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane |
| Yellow | |
| MFCD13184962 | |
| O-(2-Azidoethyl)-O′C-methylheptaethylene Glycol, mPEG8-Azide, mPEG8-N3, Methyl-PEG8-Azide | |
| COCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-] | |
| 409.48 | |
| 409.48 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator