missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ 24 LEB Selective Supplement

Catalog No. SR0243E
Change view
Click to view available options
24 LEB Selective Supplement
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SR0243E 24 LEB Selective Supplement
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SR0243E Supplier Thermo Scientific™ Supplier No. SR0243E

The 24 LEB selective supplement from Oxoid includes ten vials. One vial is required for 500 mL of medium.

  • Selectively enrich Listeria monocytogenes and other Listeria spp. from food and environmental samples
  • Medium provides optimal growing conditions for all Listeria species for PCR applications
  • Specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria

Directions:

  • Completely dissolve 21.75 g of 24 LEB base per 500 mL of distilled water
  • Sterilize by autoclaving at 121 °C for 15 min.
  • Cool to 50 °C
  • Aseptically add contents of one vial of 24 LEB selective supplement per 500 mL medium, reconstituted as directed
  • Cool to room temp before use, once made, store medium at 2 to 8 °C in dark

Technique:

  • Refer to BAX™ system user guide and product inserts for PCR Assay L. monocytogenes 24E (QB8125C) and BAX™ system PCR Assay Genus Listeria 24E (QB8135C) for instructions for use

Specifications

Appearance Yellow/orange pellet
Certifications/Compliance Adheres to ISO 11290-1:1997 standards
Content And Storage Room Temperature
Manufacturing Origin United Kingdom
Media Type Dehydrated culture media
For Use With (Application) Selective enrichment of Listeria sp from food and environmental samples
Packaging Type Tub
Quantity 10 Pack
Species Listeria