missing translation for 'onlineSavingsMsg'
Learn More

22-Tricosenoic Acid 97.0+%, TCI America™

Catalog No. T1211100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1211100MG 100 mg
T12111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1211100MG Supplier TCI America Supplier No. T1211100MG

Chemical Identifiers

CAS 65119-95-1
Molecular Formula C23H44O2
Molecular Weight (g/mol) 352.603
MDL Number MFCD00060118
InChI Key YGTSVJQQDISEHZ-UHFFFAOYSA-N
PubChem CID 543855
ChEBI CHEBI:73736
IUPAC Name tricos-22-enoic acid
SMILES C=CCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 76°C
Color White
Quantity 100 mg
Formula Weight 352.60
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 22-Tricosenoic Acid
EINECSNumber (2)-0609
TSCA No