accessibility menu, dialog, popup

UserName

H2COOH

CAS RN 65119-95-1

IUPAC Name: tricos-22-enoic acid
Synonyms: tricos-22-enoic acid
Molecular Weight (g/mol): 352.6
Molecular Formula: C23H44O2
InChi Key: YGTSVJQQDISEHZ-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCCCCCCCCCC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results