Learn More
Justrite™ 22 Gallon Sure-Grip™ EX Slimline Flammable Safety Cabinet with Manual-Closing Door
GREENER_CHOICE
Catalog No. 19822406
Change view
Click to view available options
Color:
Red
Catalog No. Color
19822406 Red
1 options
Catalog No. 19822406 Supplier Justrite™ Supplier No. 892201
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 22 Gallon Sure-Grip™ EX Slimline Flammable Safety Cabinet with Manual-Closing Door is a useful tool to help follow company-established color coding practices for flammable storage.
Use alone or alongside larger cabinets for expanded storage when floor space is limited. The standard slimline cabinet is 18 in. (457-mm) deep and fits flush alongside a 45-gallon (170-L) cabinet. The deep slimline cabinet is 34 in. (864-mm) deep and fits alongside a 60- or 90-gallon (227- or 341-L) cabinet. Both cabinets measure only 23-1/4 in. (591-mm) wide. These cabinets have all the great features and benefits of our classic cabinets as outlined below.
Specifications
Dimensions (D x W x H) 18 x 23.25 x 65 in.
Load Bearing Capacity 22 gal.
Color Red
Description Cold-rolled Steel (18 ga)
Door Style Manual-close
Material Steel (CR)