missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 20-Hydroxyecdysone ELISA Kit

Catalog No. EIAHYD
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAHYD 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAHYD Supplier Invitrogen™ Supplier No. EIAHYD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

The 20-Hydroxyecdysone Competitive ELISA quantitates 20-Hydroxyecdysone (20E) in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The 20-Hydroxyecdysone Competitive ELISA research-use-only kit is designed to quantitatively measure 20-Hydroxyecdysone independent of species. A 20-Hydroxyecdysone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit 20-Hydroxyecdysone antibodies and 20-Hydroxyecdysone-peroxidase conjugate is added to the wells. The 20-Hydroxyecdysone-peroxidase conjugate and any 20-Hydroxyecdysone in the sample will compete to bind to the anti-rabbit 20-Hydroxyecdysone antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound 20-Hydroxyecdysone-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of 20-Hydroxyecdysone present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

20-Hydroxyecdysone is the most widely occurring ecdysteroid in both plant and animal species. It controls cell death during metamorphosis of Drosophila melanogaster.

Specifications

Accession Number 20 E; C27H44O7; ecdysterone
Assay Range 102.4-25000 pg/mL
Assay Sensitivity 197.8 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 20E; C27H44O7; ecdysterone20E; C27H44O7; ecdysterone;
Interassay CV 9%
Intraassay CV 10%
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target 20-Hydroxyecdysone
Test Time 45 min
Total Assay Time 2 hr 30 min
Show More Show Less