missing translation for 'onlineSavingsMsg'
Learn More

20-Hydroxycholesterol, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111014414
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014414 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014414 Supplier Merck Emd Millipore Supplier No. H63785MG
Only null left

Specifications

Packaging Glass Bottle
CAS 516-72-3
Physical Form Neat
Linear Formula C27H46O2
MDL Number MFCD16661190
Synonym 5-Cholestene-3 beta,20 alpha-diol
Type Analytical Standard
Quantity 5 mg
Formula Weight 402.65
Percent Purity ≥98%
Grade Analytical Standard
Show More Show Less